Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199378.3 | NP_955410.1 | 1 - 2 | 190 | 86 | |
GenBank mRNA | AB125856.1 | - | 1 - 2 | 70 | 86 | |
BC067246.1 | - | 1 - 2 | 194 | 6889499 | 86 | |
FQ212332.1 | - | 224 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |