Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022962.1 | NP_075251.1 | 22 - 23 | 3944 | 115 | |
GenBank mRNA | AF081144.1 | - | 21 - 22 | 3929 | 115 | |
AF081145.1 | - | 20 - 21 | 4061 | 115 | ||
AF081146.1 | - | 22 - 23 | 3944 | 115 | ||
AF081147.1 | - | 21 - 22 | 4076 | 115 | ||
AF111099.1 | - | 23 - 24 | 4270 | 115 | ||
U72487.1 | - | 21 - 22 | 3746 | 115 | ||
U78105.1 | - | 21 - 22 | 4071 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |