Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033650.1 | NP_387499.1 | 3 - 4 | 335 | 62 | |
XM_008772474.1 | XP_008770696.1 | 2 - 3 | 1036 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |