Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012988.2 | NP_037120.1 | 8 - 9 | 1576 | 98 | |
GenBank mRNA | AB060652.1 | - | 1484 | 98 | ||
AF112455.1 | - | 1656 | 98 | |||
D78017.1 | - | 8 - 9 | 1576 | 98 | ||
D78018.1 | - | 7 - 8 | 1505 | 98 | ||
D78019.1 | - | 4 - 5 | 1169 | 98 | ||
X13167.1 | - | 8 - 9 | 1418 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |