Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271106.1 | NP_001258035.1 | 5 - 6 | 1693 | 66 | |
GenBank mRNA | AJ535460.1 | - | 5 - 6 | 1669 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Ion Channel 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |