Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106103.1 | NP_001099573.1 | 2 - 3 | 332 | 177 | |
XM_006253696.2 | XP_006253758.2 | 3 - 4 | 778 | 177 | ||
XM_006253699.2 | XP_006253761.1 | 3 - 4 | 778 | 177 | ||
XM_008771490.1 | XP_008769712.1 | 3 - 4 | 778 | 177 | ||
XM_008771558.1 | XP_008769780.1 | 3 - 4 | 778 | 177 | ||
GenBank mRNA | FQ213759.1 | - | 1 - 2 | 244 | 177 | |
FQ221052.1 | - | 1 - 2 | 244 | 177 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |