Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025730.2 | NP_001020901.1 | 5 - 6 | 800 | 60 | |
XM_006233623.2 | XP_006233685.1 | 4 - 5 | 1159 | 60 | ||
XM_006233624.2 | XP_006233686.1 | 6 - 7 | 862 | 60 | ||
XM_006233625.2 | XP_006233687.1 | 6 - 7 | 1104 | 60 | ||
XM_006233626.2 | XP_006233688.1 | 5 - 6 | 756 | 60 | ||
XM_006233627.2 | XP_006233689.1 | 5 - 6 | 1000 | 60 | ||
XM_006233628.2 | XP_006233690.1 | 4 - 5 | 516 | 60 | ||
GenBank mRNA | BC099179.1 | - | 5 - 6 | 803 | 7383052 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |