Assay Details
Gene Symbol: Prrx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105739.1 | NP_001099209.1 | 3 - 4 | 771 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P655047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101193 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P655047 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P655047 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P655047 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P655047 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100530 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92536 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58010 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10835259 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835260 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835258 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10835257 | - | - |
Rat Genome 230 2.0 Array | 1374477_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374477_at | - | - |
Rat Genome 230 2.0 Array | 1390772_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390772_at | - | - |
Affymetrix HT Rat Focus Array | 1374477_at | - | - |
Affymetrix HT Rat Focus Array | 1374477_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300002588.rn.1 | TC0300000226.rn.1 | RTA TC |
Rat Expression Set 230 | 1374477_PM_at | - | - |
Rat Expression Set 230 | 1374477_at | - | - |
Rat Expression Set 230 | 1390772_PM_at | - | - |
Rat Expression Set 230 | 1390772_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10835257 | - | - |
Rat Genome U34 Set | rc_AA944184_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21745920 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358593 | - | - |
