Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013080.2 | NP_001013098.2 | 3 - 4 | 279 | 75 | |
XM_006233590.2 | XP_006233652.1 | 3 - 4 | 261 | 75 | ||
XM_008761584.1 | XP_008759806.1 | 2 - 3 | 967 | 75 | ||
XM_008761585.1 | XP_008759807.1 | 3 - 4 | 922 | 75 | ||
GenBank mRNA | CF115110.1 | - | 3 - 4 | 279 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |