Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238143.2 | XP_006238205.1 | 5 - 6 | 1914 | 108 | |
XM_006238144.2 | XP_006238206.1 | 5 - 6 | 1867 | 108 | ||
XM_008763690.1 | XP_008761912.1 | 5 - 6 | 622 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |