Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107938.1 | NP_001101408.1 | 9 - 10 | 962 | 82 | |
XM_008763673.1 | XP_008761895.1 | 9 - 10 | 1294 | 82 | ||
GenBank mRNA | BC158699.1 | - | 9 - 10 | 950 | 9028317 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |