Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012190.1 | NP_001012190.1 | 2 - 3 | 243 | 116 | |
XM_008763626.1 | XP_008761848.1 | 2 - 3 | 252 | 116 | ||
GenBank mRNA | BC083594.1 | - | 2 - 3 | 243 | 7132783 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |