Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173098.1 | NP_775121.1 | 2 - 3 | 1180 | 95 | |
XM_006244738.2 | XP_006244800.1 | 2 - 3 | 1346 | 95 | ||
XM_008766988.1 | XP_008765210.1 | 2 - 3 | 1346 | 95 | ||
XM_008766989.1 | XP_008765211.1 | 3 - 4 | 1249 | 95 | ||
XM_008766990.1 | XP_008765212.1 | 3 - 4 | 1122 | 95 | ||
XM_008766991.1 | XP_008765213.1 | 2 - 3 | 469 | 95 | ||
GenBank mRNA | M85301.1 | - | 2 - 3 | 1180 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |