Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106914.1 | NP_001100384.1 | 5 - 6 | 3951 | 144 | |
XM_006244965.2 | XP_006245027.1 | 3 - 4 | 2000 | 144 | ||
XM_006244966.2 | XP_006245028.1 | 3 - 4 | 552 | 144 | ||
XM_006244967.2 | XP_006245029.1 | 3 - 4 | 1064 | 144 | ||
XM_008767084.1 | XP_008765306.1 | 2 - 3 | 2746 | 144 | ||
XM_008767085.1 | XP_008765307.1 | 2 - 3 | 2864 | 144 | ||
XM_008767086.1 | XP_008765308.1 | 3 - 4 | 557 | 144 | ||
XM_008767087.1 | XP_008765309.1 | 3 - 4 | 486 | 144 | ||
GenBank mRNA | FQ228496.1 | - | 4 - 5 | 1120 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |