Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108213.1 | NP_001101683.1 | 4 - 5 | 514 | 87 | |
XM_006244819.2 | XP_006244881.1 | 5 - 6 | 1246 | 87 | ||
XM_006244820.2 | XP_006244882.1 | 5 - 6 | 1866 | 87 | ||
XM_006244821.2 | XP_006244883.1 | 4 - 5 | 1048 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |