Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198789.2 | NP_942084.1 | 2 - 3 | 158 | 117 | |
XM_006245000.2 | XP_006245062.2 | 2 - 3 | 157 | 117 | ||
GenBank mRNA | BC061580.1 | - | 2 - 3 | 162 | 6890414 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |