Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gulp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013171.2 | NP_001013189.1 | 5 - 6 | 367 | 153 | |
XM_006244804.2 | XP_006244866.1 | 4 - 5 | 309 | 153 | ||
XM_008767023.1 | XP_008765245.1 | 4 - 5 | 311 | 153 | ||
XM_008767024.1 | XP_008765246.1 | 4 - 5 | 309 | 153 | ||
GenBank mRNA | BC087053.1 | - | 5 - 6 | 367 | 7123670 | 153 |
DQ668344.1 | - | 5 - 6 | 351 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |