Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107837.1 | NP_001101307.1 | 3 - 4 | 383 | 63 | |
XM_006234066.2 | XP_006234128.1 | 3 - 4 | 584 | 63 | ||
XM_008761753.1 | XP_008759975.1 | 3 - 4 | 645 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |