Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5kl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749451.3 | XP_003749499.2 | 4 - 5 | 2070 | 62 | |
XM_006234011.2 | XP_006234073.1 | 3 - 4 | 2016 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |