Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6galnac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749452.3 | XP_003749500.2 | 1 - 2 | 174 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |