Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100706.1 | NP_001094176.1 | 3 - 4 | 885 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |