Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147139.2 | NP_671480.2 | 7 - 8 | 1082 | 142 | |
GenBank mRNA | CO405730.1 | - | 4 - 5 | 486 | 142 | |
EV769502.1 | - | 3 - 4 | 413 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |