Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107923.2 | NP_001101393.1 | 1 - 2 | 168 | 75 | |
XM_006237990.2 | XP_006238052.1 | 1 - 2 | 182 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |