Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2j1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106642.1 | NP_001100112.1 | 2 - 3 | 135 | 114 | |
XM_008763612.1 | XP_008761834.1 | 2 - 3 | 233 | 114 | ||
XM_008763613.1 | XP_008761835.1 | 2 - 3 | 233 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |