Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024342.1 | NP_001019513.1 | 9 - 10 | 943 | 71 | |
GenBank mRNA | BC083662.1 | - | 9 - 10 | 943 | 7135392 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |