Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107917.2 | NP_001101387.2 | 4 - 5 | 461 | 133 | |
XM_006237932.1 | XP_006237994.1 | 5 - 6 | 583 | 133 | ||
GenBank mRNA | AM746508.1 | - | 4 - 5 | 461 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |