Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106217.1 | NP_001099687.1 | 3 - 4 | 947 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |