Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khdrbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133318.1 | NP_579852.1 | 7 - 8 | 1051 | 142 | |
XM_008766952.1 | XP_008765174.1 | 7 - 8 | 1071 | 142 | ||
GenBank mRNA | AF305618.1 | - | 7 - 8 | 1051 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |