Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106216.1 | NP_001099686.1 | 1 | 735 | 83 | |
XM_006227988.2 | XP_006228050.1 | 2 | 922 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |