Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024795.1 | NP_001019966.1 | 3 - 4 | 514 | 156 | |
XM_006253633.2 | XP_006253695.1 | 3 - 4 | 1508 | 156 | ||
XM_006253634.2 | XP_006253696.1 | 3 - 4 | 501 | 156 | ||
XM_006253637.2 | XP_006253699.1 | 3 - 4 | 2030 | 156 | ||
XM_006253641.2 | XP_006253703.1 | 3 - 4 | 1444 | 156 | ||
XM_006253642.2 | XP_006253704.1 | 3 - 4 | 1966 | 156 | ||
XM_006253644.2 | XP_006253706.1 | 3 - 4 | 437 | 156 | ||
XM_008771498.1 | XP_008769720.1 | 3 - 4 | 2030 | 156 | ||
GenBank mRNA | AY724486.1 | - | 3 - 4 | 439 | 156 | |
BC098030.1 | - | 3 - 4 | 514 | 7456296 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |