Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011966.1 | NP_001011966.1 | 13 - 14 | 1498 | 83 | |
XM_006235888.2 | XP_006235950.1 | 12 - 13 | 1595 | 83 | ||
XM_006235889.2 | XP_006235951.1 | 12 - 13 | 1559 | 83 | ||
GenBank mRNA | BC085349.1 | - | 13 - 14 | 1498 | 7107899 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |