Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025155.2 | NP_001020326.1 | 2 - 3 | 1096 | 63 | |
XM_008762657.1 | XP_008760879.1 | 2 - 3 | 1384 | 63 | ||
GenBank mRNA | BC079098.1 | - | 2 - 3 | 973 | 7124390 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |