Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107222.1 | NP_001100692.1 | 4 - 5 | 566 | 89 | |
XM_006251076.2 | XP_006251138.1 | 4 - 5 | 647 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |