Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108363.1 | NP_001101833.1 | 1 - 2 | 385 | 89 | |
XM_006251147.2 | XP_006251209.1 | 1 - 2 | 653 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |