Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bod1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770238.1 | XP_008768460.1 | 5 - 6 | 1442 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |