Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105750.2 | NP_001099220.2 | 9 - 10 | 1465 | 84 | |
XM_008770187.1 | XP_008768409.1 | 9 - 10 | 8107 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |