Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032080.1 | NP_114469.1 | 5 - 6 | 746 | 139 | |
XM_006248373.2 | XP_006248435.1 | 5 - 6 | 1452 | 139 | ||
XM_006248374.2 | XP_006248436.1 | 5 - 6 | 1452 | 139 | ||
XM_006248375.2 | XP_006248437.1 | 5 - 6 | 1452 | 139 | ||
XM_008768745.1 | XP_008766967.1 | 5 - 6 | 1452 | 139 | ||
GenBank mRNA | X53428.1 | - | 5 - 6 | 721 | 139 | |
X73653.1 | - | 5 - 6 | 746 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |