Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005549.1 | NP_001005549.1 | 8 - 9 | 1179 | 154 | |
GenBank mRNA | BC081887.1 | - | 8 - 9 | 1179 | 7109073 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |