Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcaim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110838.1 | NP_001104308.1 | 6 - 7 | 810 | 120 | |
XM_006244192.2 | XP_006244254.1 | 6 - 7 | 922 | 120 | ||
XM_006244193.2 | XP_006244255.1 | 6 - 7 | 1031 | 120 | ||
XM_006244194.2 | XP_006244256.1 | 6 - 7 | 947 | 120 | ||
XM_006244195.2 | XP_006244257.1 | 6 - 7 | 915 | 120 | ||
XM_008766717.1 | XP_008764939.1 | 6 - 7 | 947 | 120 | ||
XM_008766718.1 | XP_008764940.1 | 5 - 6 | 1056 | 120 | ||
XM_008766719.1 | XP_008764941.1 | 5 - 6 | 814 | 120 | ||
XM_008766720.1 | XP_008764942.1 | 5 - 6 | 779 | 120 | ||
XM_008766721.1 | XP_008764943.1 | 5 - 6 | 811 | 120 | ||
XM_008766722.1 | XP_008764944.1 | 6 - 7 | 921 | 120 | ||
XM_008766723.1 | XP_008764945.1 | 5 - 6 | 926 | 120 | ||
XM_008766724.1 | XP_008764946.1 | 6 - 7 | 839 | 120 | ||
XM_008766725.1 | XP_008764947.1 | 6 - 7 | 843 | 120 | ||
XM_008766726.1 | XP_008764948.1 | 5 - 6 | 707 | 120 | ||
GenBank mRNA | FQ210480.1 | - | 6 - 7 | 824 | 120 | |
FQ211645.1 | - | 6 - 7 | 895 | 120 | ||
FQ215070.1 | - | 6 - 7 | 824 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |