Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191819.1 | NP_001178748.1 | 3 - 4 | 537 | 66 | |
XM_006243962.1 | XP_006244024.1 | 3 - 4 | 520 | 66 | ||
XM_006243963.2 | XP_006244025.1 | 3 - 4 | 520 | 66 | ||
XM_008766630.1 | XP_008764852.1 | 4 - 5 | 569 | 66 | ||
XM_008766631.1 | XP_008764853.1 | 3 - 4 | 520 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |