Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191884.1 | NP_001178813.1 | 3 - 4 | 311 | 84 | |
XM_006243847.2 | XP_006243909.1 | 3 - 4 | 320 | 84 | ||
XM_006243848.2 | XP_006243910.1 | 3 - 4 | 246 | 84 | ||
XM_006243849.1 | XP_006243911.1 | 3 - 4 | 764 | 84 | ||
XM_006243850.2 | XP_006243912.1 | 3 - 4 | 285 | 84 | ||
XM_006243851.1 | XP_006243913.1 | 3 - 4 | 349 | 84 | ||
GenBank mRNA | AF051943.1 | - | 1 - 2 | 53 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |