Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mei4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109378.1 | NP_001102848.1 | 1 - 2 | 139 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |