Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104528.1 | NP_001097998.1 | 3 - 4 | 1072 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Rattus norvegicus Probe spans exons |