Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191691.2 | NP_001178620.2 | 8 - 9 | 2136 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |