Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrfap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009264.1 | NP_001009264.1 | 1 - 2 | 511 | 70 | |
GenBank mRNA | AF526268.1 | - | 2 - 3 | 461 | 70 | |
BC088308.1 | - | 1 - 2 | 511 | 7324285 | 70 | |
FQ228750.1 | - | 1 - 2 | 531 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |