Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cytip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012086.1 | NP_001012086.1 | 3 - 4 | 323 | 109 | |
XM_006234202.2 | XP_006234264.1 | 4 - 5 | 297 | 109 | ||
GenBank mRNA | BC088207.1 | - | 3 - 4 | 323 | 7381695 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |