Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cytip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012086.1 | NP_001012086.1 | 6 - 7 | 594 | 69 | |
XM_006234202.2 | XP_006234264.1 | 7 - 8 | 568 | 69 | ||
GenBank mRNA | BC088207.1 | - | 6 - 7 | 594 | 7381695 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |