Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgef1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728083.4 | XP_002728129.2 | 2 - 3 | 350 | 119 | |
XM_008770077.1 | XP_008768299.1 | 2 - 3 | 350 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |