Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004275.1 | NP_001004275.1 | 2 - 3 | 193 | 76 | |
XM_006250668.2 | XP_006250730.1 | 2 - 3 | 239 | 76 | ||
GenBank mRNA | BC079384.1 | - | 2 - 3 | 193 | 7111425 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |