Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004275.1 | NP_001004275.1 | 5 - 6 | 605 | 129 | |
XM_006250667.2 | XP_006250729.1 | 4 - 5 | 515 | 129 | ||
XM_006250668.2 | XP_006250730.1 | 5 - 6 | 651 | 129 | ||
GenBank mRNA | BC079384.1 | - | 5 - 6 | 605 | 7111425 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |